Learn More
Thermo Scientific Chemicals L-Serine, Cell Culture Reagent
CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol
Supplier: Thermo Scientific Chemicals J6218736
| Quantity | 500 g |
|---|
Description
It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.
Solubility
Soluble in water.
Notes
Store in cool, dry place. Incompatible with oxidizing agents.
Chemical Identifiers
| C3H7NO3 | |
| MFCD00064224 | |
| l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
| CHEBI:17115 | |
| C(C(C(=O)O)N)O |
Specifications
| 222°C | |
| 56-45-1 | |
| C3H7NO3 | |
| 1721404 | |
| l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid | |
| MTCFGRXMJLQNBG-REOHCLBHSA-N | |
| (2S)-2-amino-3-hydroxypropanoic acid | |
| 105.093 | |
| CHEBI:17115 | |
| Cell Culture Reagent |
| L-Serine | |
| White | |
| MFCD00064224 | |
| 14,8460 | |
| Soluble in water. | |
| C(C(C(=O)O)N)O | |
| 500 g | |
| 5951 | |
| 105.09 | |
| Powder |
Safety and Handling
EINECSNumber : 200-274-3
RTECSNumber : VT8100000
TSCA : Yes
Recommended Storage : Ambient temperatures