missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Serine, Cell Culture Reagent

Catalog No. AAJ6218709
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6218709 10 g
AAJ6218722 100 g
AAJ6218736 500 g
AAJ62187A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAJ6218709 Supplier Thermo Scientific Chemicals Supplier No. J6218709

CAS: 56-45-1 | C3H7NO3 | 105.093 g/mol

It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a precursor of glycine by serine hydroxymethyltransferase. D-serine, synthesized by serine racemase from L-serine, serves as a neuronal signaling molecule by activating NMDA receptors in the brain.

Solubility
Soluble in water.

Notes
Store in cool, dry place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 56-45-1
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.093
MDL Number MFCD00064224
InChI Key MTCFGRXMJLQNBG-REOHCLBHSA-N
Synonym l-serine, serine, s-2-amino-3-hydroxypropanoic acid, s-serine, beta-hydroxyalanine, h-ser-oh, l-ser, l---serine, serine, l, 2s-2-amino-3-hydroxypropanoic acid
PubChem CID 5951
ChEBI CHEBI:17115
IUPAC Name (2S)-2-amino-3-hydroxypropanoic acid
SMILES C(C(C(=O)O)N)O

Specifications

Melting Point 222°C
Color White
Beilstein 1721404
Merck Index 14,8460
Solubility Information Soluble in water.
Quantity 10 g
Formula Weight 105.09
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Serine
EINECSNumber 200-274-3
RTECSNumber VT8100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only