missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ L-Selenomethionine, Calbiochem™,
$396.00 - $808.00
Chemical Identifiers
| CAS | 3211-76-5 |
|---|---|
| Molecular Formula | C5H11NO2Se |
| Molecular Weight (g/mol) | 196.12 |
| MDL Number | MFCD00037210 |
| InChI Key | RJFAYQIBOAGBLC-BYPYZUCNSA-N |
| Synonym | l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid |
| PubChem CID | 105024 |
| ChEBI | CHEBI:62621 |
| IUPAC Name | (2S)-2-amino-4-(methylselanyl)butanoic acid |
| SMILES | C[Se]CC[C@H](N)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
561505250MG
|
MilliporeSigma™
561505250MG |
250 mg | Glass Bottle |
Each for $396.00
|
|
||||
|
5615051GM
|
MilliporeSigma™
5615051GM |
1 g | Glass Bottle |
Each for $808.00
|
|
||||
Chemical Identifiers
| C5H11NO2Se | |
| MFCD00037210 | |
| l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid | |
| CHEBI:62621 | |
| C[Se]CC[C@H](N)C(O)=O |
Specifications
| 3211-76-5 | |
| C5H11NO2Se | |
| l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid | |
| C[Se]CC[C@H](N)C(O)=O | |
| 196.12 | |
| CHEBI:62621 | |
| Solid |
| White | |
| MFCD00037210 | |
| RJFAYQIBOAGBLC-BYPYZUCNSA-N | |
| (2S)-2-amino-4-(methylselanyl)butanoic acid | |
| 105024 | |
| ≥98% (HPLC) |