missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ L-Selenomethionine, Calbiochem™,
Supplier: MilliporeSigma™ 5615051GM
| Quantity | 1 g |
|---|---|
| Packaging | Glass Bottle |
Chemical Identifiers
| C5H11NO2Se | |
| MFCD00037210 | |
| l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid | |
| CHEBI:62621 | |
| C[Se]CC[C@H](N)C(O)=O |
Specifications
| 3211-76-5 | |
| C5H11NO2Se | |
| MFCD00037210 | |
| RJFAYQIBOAGBLC-BYPYZUCNSA-N | |
| (2S)-2-amino-4-(methylselanyl)butanoic acid | |
| 1 g | |
| CHEBI:62621 | |
| Solid |
| White | |
| Glass Bottle | |
| l-selenomethionine, l-+-selenomethionine, selenium-l-methionine, l-selenomethioninum, seleno-l-methionine, s-2-amino-4-methylselanyl butanoic acid, semet, s-2-amino-4-methylseleno butyric acid, unii-964mrk2pel, s-2-amino-4-methylseleno butanoic acid | |
| C[Se]CC[C@H](N)C(O)=O | |
| 196.12 | |
| 105024 | |
| ≥98% (HPLC) |