missing translation for 'onlineSavingsMsg'
Learn More

L-Prolinol 97.0+%, TCI America™

Catalog No. P10875G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P10875G 5 g
P108725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P10875G Supplier TCI America Supplier No. P10875G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23356-96-9
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00005255
InChI Key HVVNJUAVDAZWCB-YFKPBYRVSA-N
Synonym l-prolinol, l-+-prolinol, s-pyrrolidin-2-ylmethanol, s-+-2-pyrrolidinemethanol, s-prolinol, 2s-pyrrolidin-2-ylmethanol, l +-prolinol, 2s-pyrrolidin-2-yl methanol, s-+-2-hydroxymethyl pyrrolidine, s-+-prolinol
PubChem CID 640091
IUPAC Name [(2S)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)CO

Specifications

Color Yellow
Boiling Point 66°C
Quantity 5 g
Formula Weight 101.15
Percent Purity ≥97.0% (GC,T)
Grade Not Available
Physical Form Liquid
Chemical Name or Material L-Prolinol