missing translation for 'onlineSavingsMsg'
Learn More

L-Phenylalanine, 98.5%, Spectrum™ Chemical

Chemical Identifiers

CAS 63-91-2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
18611380
View Documents Promotion Details
N/A 25 g Poly Pail N/A
Only null left
 
18611379
View Documents Promotion Details
N/A 100 g Poly Pail N/A
Only null left
 
18611378
View Documents Promotion Details
N/A 1 kg Poly Pail N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

63-91-2

Specifications

63-91-2
1
10 ppm
-33.2° to -35.2°
Reagent
1.5 ppm
20 ppm
0.003
98.5%
L-Phenylalanine
Videos
SDS
Documents

Documents

CAUTION: For manufacturing, processing or repacking. Read and understand the label and Safety Data Sheet (SDS) prior to use.