missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Phenylalanine, 98.5%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor PH145100GM
| Quantity | 100 g |
|---|---|
| Packaging | Poly Pail |
Chemical Identifiers
| 63-91-2 |
Specifications
| L-Phenylalanine | |
| 1.5 ppm | |
| 20 ppm | |
| 0.003 | |
| -33.2° to -35.2° | |
| 98.5% |
| 63-91-2 | |
| 1 | |
| 10 ppm | |
| Poly Pail | |
| 100 g | |
| Reagent |