missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Methionine sulfone, 98+%

Catalog No. AAA1702703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1702703 1 g
AAA1702706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1702703 Supplier Thermo Scientific Chemicals Supplier No. A1702703

CAS: 7314-32-1 | C5H11NO4S | 181.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7314-32-1
Molecular Formula C5H11NO4S
Molecular Weight (g/mol) 181.21
MDL Number MFCD00066020
InChI Key UCUNFLYVYCGDHP-BYPYZUCNSA-N
Synonym l-methionine sulfone, s-2-amino-4-methylsulfonyl butanoic acid, s-dioxymethionine, l-2-amino-4-methylsulfonyl butanoic acid, 2s-2-amino-4-methylsulfonylbutanoic acid, 2s-2-amino-4-methanesulfonylbutanoic acid, methionine s,s-dioxide, s-amino-4-methylsulphonyl butyric acid, h-met o 2-oh, l-methioninesulfone,99+%
PubChem CID 445282
ChEBI CHEBI:21363
IUPAC Name (2S)-2-amino-4-methylsulfonylbutanoic acid
SMILES CS(=O)(=O)CC[C@H](N)C(O)=O

Specifications

Melting Point 254°C (decomposition)
Beilstein 1725510
Quantity 1 g
Formula Weight 181.21
Optical Rotation +11.6° (c=0.69 in Water)
Percent Purity ≥99%
Chemical Name or Material L-Methionine sulfone
EINECSNumber 230-774-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only