missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ L-Methionine, 98+%
Supplier: Thermo Scientific™ 166161000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécifications
| L-Methionine | |
| 300ppm max. | |
| 500ppm max. | |
| 0.5% max. of any individual impurity | |
| 30ppm max. | |
| 98+% | |
| Plastic bottle | |
| MFCD00063097 | |
| 04, II, 938 | |
| 15, 6047 | |
| 0.1% max. | |
| CSCCC(C(=O)O)N | |
| +23.25° (20°C c=2,6N HCl) | |
| 100g | |
| CHEBI:16643 | |
| ≥98% |
| 98+% | |
| 63-68-3 | |
| 15ppm max. | |
| Authentic | |
| 0.5% max. (1 g, 105°C) | |
| C5H11NO2S | |
| CH3SCH2CH2CH(NH2)CO2H | |
| + 23.25 | |
| 300ppm max. | |
| l-methionine, methionine, h-met-oh, s-2-amino-4-methylthio butanoic acid, cymethion, l---methionine, liquimeth, s-methionine, l-methioninum, methilanin | |
| FFEARJCKVFRZRR-BYPYZUCNSA-N | |
| (2S)-2-amino-4-methylsulfanylbutanoic acid | |
| 149.208 | |
| 6137 | |
| 149.21 |
Sécurité et manipulation
AIR SENSITIVE
EINECSNumber : 200-562-9
RTECSNumber : PD0457000
TSCA : TSCA