missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Mannose, 99%

CAS: 10030-80-5 | C6H12O6 | 180.16 g/mol

Supplier:  Thermo Scientific Chemicals A1772203

 View more versions of this product

Catalog No. AAA1772203


Only null left

Description

Description

L-Mannose is used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Mannose is used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.

Solubility
Soluble in Water (100 mg/mL).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

L-(-)-Mannose
129°C to 131°C
MFCD00136021
1724628
Soluble in Water (100mg/mL).
OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O
180.16
CHEBI:37681
99%
10030-80-5
C6H12O6
1 g
l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
GZCGUPFRVQAUEE-BXKVDMCESA-N
(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
82308
180.16

Chemical Identifiers

10030-80-5
180.16
GZCGUPFRVQAUEE-BXKVDMCESA-N
82308
(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
C6H12O6
MFCD00136021
l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
CHEBI:37681
OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-080-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only