missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(-)-Mannose, 99%

CAS: 10030-80-5 | C6H12O6 | 180.16 g/mol

$95.40 - $214.14

Chemical Identifiers

CAS 10030-80-5
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00136021
InChI Key GZCGUPFRVQAUEE-BXKVDMCESA-N
Synonym l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
PubChem CID 82308
ChEBI CHEBI:37681
IUPAC Name (2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAA1772201
View Documents Promotion Details
Thermo Scientific Chemicals
A1772201
0.25 g
Each for $95.40
Only null left
 
AAA1772203
View Documents Promotion Details
Thermo Scientific Chemicals
A1772203
1 g
Each for $214.14
Only null left
 
Description

Description

L-Mannose is used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Mannose is used in a study to assess substrate specificity of galactokinase from Streptococcus pneumoniae. It has also been used in a study to investigate a versatile route to L-hexoses.

Solubility
Soluble in Water (100 mg/mL).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

10030-80-5
180.16
GZCGUPFRVQAUEE-BXKVDMCESA-N
82308
(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
C6H12O6
MFCD00136021
l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
CHEBI:37681
OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O

Specifications

10030-80-5
C6H12O6
1724628
Soluble in Water (100mg/mL).
OC[C@H](O)[C@H](O)[C@@H](O)[C@@H](O)C=O
180.16
CHEBI:37681
99%
129°C to 131°C
MFCD00136021
l-mannose, l---mannose, aldehydo-l-mannose, mannose, l, 2r,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, unii-2w3ye50tx8, aldehydo-l-manno-hexose, l-?-mannose
GZCGUPFRVQAUEE-BXKVDMCESA-N
(2R,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
82308
180.16
L-(-)-Mannose
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 233-080-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only