missing translation for 'onlineSavingsMsg'
Learn More

L-Malic acid, ≥99%

Catalog No. AAJ1870036
Change view
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ1870036 500 g Plastic bottle
AAJ18700A1 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ1870036 Supplier Thermo Fisher Scientific Supplier No. J1870036

CAS: 97-67-6 | C4H6O5 | 134.087 g/mol

Chemical Identifiers

CAS 97-67-6
Molecular Formula C4H6O5
Molecular Weight (g/mol) 134.087
MDL Number MFCD00064213
InChI Key BJEPYKJPYRNKOW-REOHCLBHSA-N
Synonym l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2 s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
PubChem CID 222656
ChEBI CHEBI:30797
IUPAC Name (2 S)-2-hydroxybutanedioic acid
SMILES C(C(C(=O)O)O)C(=O)O

Specifications

Melting Point 100°C to 106°C
Beilstein 1723541
Formula Weight 134.09
Percent Purity ≥99%
Density 1.6
Quantity 500 g
Flash Point >110°C
Packaging Plastic bottle
Physical Form Crystalline Powder
Specific Gravity 1.6
Chemical Name or Material L-Malic acid
Show More Show Less
Hazard Category H315-H318-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
EINECSNumber 202-601-5
RTECSNumber ON7175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only