missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Lysine monohydrate, 99%

Catalog No. AC173731000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173731000 100 g Plastic Bottle
AC173730250 25 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173731000 Supplier Thermo Scientific Chemicals Supplier No. 173731000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39665-12-8 | C6H16N2O3 | 164.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
SMILES O.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 212.0°C to 214.0°C
Ammonium 200ppm max.
pH 9 to 10.5 (2.5% soln. at 25°C)
Color White to Yellow
Chlorine (Cl) 300ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Other Amino Acids 0.5% max. (TLC)
Packaging Plastic Bottle
Linear Formula H2N(CH2)4CH(NH2)CO2H·H2O
Specific Rotation + 26.25
Beilstein 04, 435
Sulfate 200ppm max.
Merck Index 14, 5636
Transmittance 95% min. (c=5, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 1500g/L (20°C). Other solubilities: insoluble in common neutral solvents
Water 10 to 11.5%
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrate
Specific Rotation Condition + 26.25 (20.00°C c=8,6N HCl)
Quantity 100 g
Formula Weight 164.21
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(+)-Lysine monohydrate
Show More Show Less

RUO – Research Use Only