missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine monohydrate, 97%

Catalog No. AAA1304036
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1304036 500 g
AAA1304014 25 g
AAA1304022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1304036 Supplier Thermo Scientific Chemicals Supplier No. A1304036
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39665-12-8 | C6H16N2O3 | 164.21 g/mol

L-Lysine is an essential proteinogenic alfa amino acid used in various applications such as non-animal sourced supplement in cell culture media. It acts as a substrate of enzymes like L-lysine oxidase as well as a component of poly-lysine polymers. It is also involved in the studies of glycation mechanism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Lysine is an essential proteinogenic alfa amino acid used in various applications such as non-animal sourced supplement in cell culture media. It acts as a substrate of enzymes like L-lysine oxidase as well as a component of poly-lysine polymers. It is also involved in the studies of glycation mechanism.

Solubility
Soluble in water and hydrochloric acid.

Notes
Hygroscopic. Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
SMILES O.NCCCC[C@H](N)C(O)=O

Specifications

Melting Point 210°C (decomposition)
Beilstein 1722531
Merck Index 14,5636
Solubility Information Soluble in water and hydrochloric acid.
IUPAC Name 2,6-diaminohexanoic acid hydrate
Quantity 500 g
Formula Weight 164.21 (146.19 Anhydrous)
Optical Rotation +22.5° (c=2 in 6M HCl)
Percent Purity 97%
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material L-Lysine monohydrate
Show More Show Less
EINECSNumber 200-294-2
RTECSNumber OL5540000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only