missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine, 99.6%, MP Biomedicals™

Catalog No. ICN19022425
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19022425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19022425 Supplier MP Biomedicals Inc Supplier No. 0219022425

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 39665-12-8
Molecular Formula C6H16N2O3
Molecular Weight (g/mol) 164.21
MDL Number MFCD00151035
InChI Key HZRUTVAFDWTKGD-XZNNNFJINA-N
Synonym l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1
PubChem CID 16211825
IUPAC Name (2S)-2,6-diaminohexanoic acid hydrate
SMILES O.NCCCC[C@H](N)C(O)=O

Specifications

Color White
Quantity 25 g
Formula Weight 164.2
Physical Form Crystalline Powder
Chemical Name or Material L-lysine, Free Base, Monohydrate