missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Lysine, 98%

Numéro de catalogue. p-3776610
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC303341000 100 g Glass Bottle
AC303345000 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AC303341000 Fournisseur Thermo Scientific Chemicals Code fournisseur 303341000
Il en reste null

CAS: 56-87-1 | C6H14N2O2 | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Identifiants chimiques

CAS 56-87-1
Molecular Formula C6H14N2O2
Molecular Weight (g/mol) 146.19
MDL Number MFCD00064433
InChI Key KDXKERNSBIXSRK-MLHKIVSYNA-N
Synonym l-lysine, lysine, h-lys-oh, lysine acid, s-lysine, aminutrin, 2s-2,6-diaminohexanoic acid, l-+-lysine, alpha-lysine, lysinum
PubChem CID 5962
ChEBI CHEBI:18019
IUPAC Name (2S)-2,6-diaminohexanoic acid
SMILES NCCCC[C@H](N)C(O)=O

Spécifications

Melting Point 214.0°C to 216.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation +23.0° to +27.0° (20°C, 589nm) (C=2, 1N HCl)
Water 3% max.
Quantity 100 g
Formula Weight 146.19
Percent Purity 98%
Physical Form Powder or Crystals
Chemical Name or Material L-Lysine
Afficher plus de résultats Afficher moins de résultats

RUO – Research Use Only