missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Leucine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 167°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 223.74 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | L-Leucine tert-Butyl Ester Hydrochloride |
Safety and Handling
| TSCA | No |