missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine Ethyl Ester Hydrochloride 99.0+%, TCI America™

Catalog No. L012325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L012325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. L012325G Supplier TCI America Supplier No. L012325G

Chemical Identifiers

CAS 2743-40-0
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00034879
InChI Key QIGLJVBIRIXQRN-ZETCQYMHSA-N
Synonym l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl
PubChem CID 11658447
IUPAC Name ethyl (2S)-2-amino-4-methylpentanoate
SMILES CCOC(=O)[C@@H](N)CC(C)C

Specifications

Melting Point 136°C
Color White
Quantity 25 g
Formula Weight 195.69
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Leucine Ethyl Ester Hydrochloride