missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine ethyl ester hydrochloride, 97%

Catalog No. AAL1314714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1314714 25 g
AAL1314706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1314714 Supplier Thermo Scientific Chemicals Supplier No. L1314714
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2743-40-0 | C8H17NO2 | 159.23 g/mol

L-Leucine ethyl ester hydrochloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Leucine ethyl ester hydrochloride is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2743-40-0
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00034879
InChI Key QIGLJVBIRIXQRN-ZETCQYMHSA-N
Synonym l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl
PubChem CID 11658447
SMILES CCOC(=O)[C@@H](N)CC(C)C

Specifications

Melting Point 134°C to 137°C
Beilstein 3994312
Solubility Information Soluble in water.
IUPAC Name ethyl (2S)-2-amino-4-methylpentanoate;hydrochloride
Quantity 25 g
Odor Odorless
Formula Weight 195.69
Optical Rotation +18° (c=5 in Ethanol)
Percent Purity 97%
Chemical Name or Material L-Leucine ethyl ester hydrochloride
EINECSNumber 220-375-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only