missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-isoleucine (cristaux blancs ou poudre cristalline), Fisher BioReagents™
Description
Cet acide aminé convient à une utilisation dans les systèmes de culture tissulaire nécessitant des additifs.
Spécifications
Spécifications
| Point de fusion | 284 °C |
| Nom ou substance chimique | L-Isoleucine |
| Ammonium | 0,02% max. |
| Arsenic (As) | 1 ppm maximum. |
| pH | 5,5 à 6,5 |
| Couleur | Blanc |
| Métaux lourds (sous forme de Pb) | 10 ppm max. |
| Résidus de calcination | 0,1% max. |
| Désignation | Test Réussi |
| Perte à la dessication | 0,2% max. |
| Afficher plus de résultats |
Sécurité et manipulation
| Hazard Statement | CAUTION! |
| Precautionary Statement | Emergency Overview May be harmful by inhalation, in contact with skin and if swallowed. Avoid contact with skin and eyes. Do not breathe dust. Do not breathe vapors or spray mist. Do not ingest. Use personal protective equipment. Wash off immediately with soap and plenty of water removing all contaminated clothes and shoes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Remove from exposure, lie down. Move to fresh air. Move to fresh air. Obtain medical attention. NFPA Health:1 Flammability:1 Instability:0 |
| Recommended Storage | RT |