missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine tert-Butyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. I06855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06855G 5 g
I06851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06855G Supplier TCI America Supplier No. I06855G

Chemical Identifiers

CAS 69320-89-4
Molecular Formula C10H22ClNO2
Molecular Weight (g/mol) 223.74
MDL Number MFCD00058015
InChI Key IFRYMHOZFAPYPJ-HAGKNZRZNA-N
Synonym h-ile-otbu.hcl, l-isoleucine tert-butyl ester hydrochloride, h-ile-otbu hcl, l-isoleucine t-butyl ester hydrochloride, h-ile-otbu hydrochloride, l-isoleucine t-butyl ester hcl, tert-butyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, l-isoleucine, 1,1-dimethylethyl ester, hydrochloride, h-ile-obut.hcl, h-ile-otbu??hcl
PubChem CID 11687228
IUPAC Name tert-butyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
SMILES Cl.CC[C@H](C)[C@H](N)C(=O)OC(C)(C)C

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 223.74
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Isoleucine tert-Butyl Ester Hydrochloride