missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine tert-butyl ester hydrochloride, 98%

Catalog No. AAH5984314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5984314 25 g
AAH5984306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5984314 Supplier Thermo Scientific Chemicals Supplier No. H5984314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 69320-89-4 | C10H22ClNO2 | 223.74 g/mol

It is used as a pharmaceutical industry. t is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical industry. t is an important raw material and intermediate used in organic synthesis, agrochemicals and dyestuffs.

Solubility
Soluble in 50 mg/ml methanol.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 69320-89-4
Molecular Formula C10H22ClNO2
Molecular Weight (g/mol) 223.74
MDL Number MFCD00058015
InChI Key IFRYMHOZFAPYPJ-HAGKNZRZNA-N
Synonym h-ile-otbu.hcl, l-isoleucine tert-butyl ester hydrochloride, h-ile-otbu hcl, l-isoleucine t-butyl ester hydrochloride, h-ile-otbu hydrochloride, l-isoleucine t-butyl ester hcl, tert-butyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, l-isoleucine, 1,1-dimethylethyl ester, hydrochloride, h-ile-obut.hcl, h-ile-otbu??hcl
PubChem CID 11687228
IUPAC Name tert-butyl (2S,3S)-2-amino-3-methylpentanoate;hydrochloride
SMILES Cl.CC[C@H](C)[C@H](N)C(=O)OC(C)(C)C

Specifications

Linear Formula C10H21NO2·HCl
Solubility Information Soluble in 50mg/ml methanol.
Quantity 25 g
Formula Weight 223.74
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Isoleucine tert-butyl ester hydrochloride
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only