missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine Methyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. I05225G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I05225G 5 g
I05221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I05225G Supplier TCI America Supplier No. I05225G

Chemical Identifiers

CAS 18598-74-8
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00038911
InChI Key GGTBEWGOPAFTTH-GEMLJDPKSA-N
Synonym l-isoleucine methyl ester hydrochloride, h-ile-ome.hcl, methyl l-isoleucinate hydrochloride, methyl l-isoleucinate hcl, methyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, 2s,3s-methyl 2-amino-3-methylpentanoate hydrochloride, l-isoleucine, methyl ester, hydrochloride, l-isoleucine methyl ester hydrochloride;h-ile-ome.hcl, h-lle-ome.hcl, h-lle-ome?cl
PubChem CID 11171351
IUPAC Name methyl (2S,3S)-2-amino-3-methylpentanoate hydrochloride
SMILES Cl.CC[C@H](C)[C@H](N)C(=O)OC

Specifications

Melting Point 98°C
Color White
Quantity 5 g
Formula Weight 181.66
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Isoleucine Methyl Ester Hydrochloride