missing translation for 'onlineSavingsMsg'
Learn More

L-Homophenylalanine 98.0+%, TCI America™

Catalog No. H09851G
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09851G 1 g
H0985100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09851G Supplier TCI America Supplier No. H09851G

Chemical Identifiers

CAS 943-73-7
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00002619
InChI Key JTTHKOPSMAVJFE-VIFPVBQESA-N
Synonym l-homophenylalanine, homophenylalanine, s-2-amino-4-phenylbutanoic acid, s-2-amino-4-phenylbutyric acid, 2s-2-amino-4-phenylbutanoic acid, homophenylalanine, l, h-hophe-oh, +-2-amino-4-phenylbutyric acid, l-homophe
PubChem CID 2724505
ChEBI CHEBI:43103
IUPAC Name (2S)-2-amino-4-phenylbutanoic acid
SMILES N[C@@H](CCC1=CC=CC=C1)C(O)=O

Specifications

Color White
Quantity 1 g
Formula Weight 179.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Homophenylalanine