missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine monohydrochloride monohydrate, Cell Culture Reagent

CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol

Supplier:  Thermo Scientific Chemicals J6146522

Quantity 100 g

 View more versions of this product

Catalog No. AAJ6146522


Only null left

Description

Description

L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

Solubility
Freely soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Chemical Identifiers

5934-29-2
209.63
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
165377
O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
C6H12ClN3O3
MFCD00151027
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
(2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride

Specifications

∽240°C (decomposition)
5934-29-2
C6H12ClN3O3
4168261
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
(2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
209.63
209.63 (191.62 Anhydrous)
Cell Culture Reagent
L-Histidine monohydrochloride monohydrate
White
MFCD00151027
14,4720
Freely soluble in water
O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
100 g
165377
+10° (c=5 in 5N HCl)
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 211-438-9

RTECSNumber : MS3119000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions