missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Histidine Monohydrochloride, Monohydrate, 98.5-101.5%, Spectrum™ Chemical
HI125, 5934-29-2, C6H9N3O2·HCl·H2O
$121.13 - $5,062.92
Chemical Identifiers
| CAS | 5934-29-2 |
|---|---|
| Molecular Formula | C6H12ClN3O3 |
| Molecular Weight (g/mol) | 209.63 |
| MDL Number | MFCD00151027 |
| InChI Key | CMXXUDSWGMGYLZ-XRIGFGBMSA-N |
| IUPAC Name | (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride |
| SMILES | O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18603848
|
N/A | 25 g | Poly Bottle |
Each for $121.13
|
|
||||
|
18603849
|
N/A | 100 g | Poly Bottle |
Each for $285.25
|
|
||||
|
18603850
|
N/A | 1 kg | Poly Bottle |
Each for $1,263.69
|
|
||||
|
18603851
|
N/A | 5 kg | Poly Pail |
Each for $5,062.92
|
|
||||
Description
Spectrum™ Chemical L-Histidine Monohydrochloride, Monohydrate , also known as 2-Amino-3-(1H-imidazol-4-yl)propanoic acid, is an α-amino acid with an imidazole functional group. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 5934-29-2 | |
| 209.63 | |
| CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
| O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O |
| C6H12ClN3O3 | |
| MFCD00151027 | |
| (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride |
Specifications
| 5934-29-2 | |
| 100% | |
| 0.1% | |
| C6H12ClN3O3 | |
| +8.5° to +10.5° | |
| O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O | |
| 209.63 | |
| Reagent |
| 0.015% | |
| 0.001% | |
| 0.3% | |
| MFCD00151027 | |
| CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
| (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride | |
| 98.5 to 101.5% |