missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine Monohydrochloride, Monohydrate, 98.5-101.5%, Spectrum™ Chemical

Catalog No. 18603848
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
Packaging:
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603848 25 g Poly Bottle
18603849 100 g Poly Bottle
18603850 1 kg Poly Bottle
18603851 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18603848 Supplier Spectrum Chemical Mfg Cor Supplier No. HI12525GM
Get Up to $500 in Chemicals with Purchase
View all promotions

HI125, 5934-29-2, C6H9N3O2·HCl·H2O

Spectrum™ Chemical L-Histidine Monohydrochloride, Monohydrate , also known as 2-Amino-3-(1H-imidazol-4-yl)propanoic acid, is an α-amino acid with an imidazole functional group. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Arsenic (As) 0.015%
CAS Max % 100%
Heavy Metals (as Pb) 0.001%
Ignition Residue 0.1%
Loss on Drying 0.3%
Packaging Poly Bottle
Specific Rotation +8.5° to +10.5°
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade Reagent