accessibility menu, dialog, popup

UserName

OHONH2(S)NHNHClOHH

CAS RN 5934-29-2

IUPAC Name: (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
Synonyms: L-histidin hydrate hydrochloride
Molecular Weight (g/mol): 209.63
Molecular Formula: C6H12ClN3O3
InChi Key: CMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILES: O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
Quantity 
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (4)
  • (4)
  • (3)
  • (4)
Grade 
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (11)
  • (4)
  • (1)
  • (4)
  • (4)
  • (10)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (4)
  • (4)
  • (4)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)