missing translation for 'onlineSavingsMsg'
Learn More

L-Gulonic acid-1,4-lactone, 95%

Catalog No. AAJ6693406
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6693406 5 g
AAJ6693414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6693406 Supplier Thermo Scientific Chemicals Supplier No. J6693406

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1128-23-0 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1128-23-0
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00064331
InChI Key SXZYCXMUPBBULW-SKNVOMKLSA-N
Synonym l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone
PubChem CID 439373
ChEBI CHEBI:17587
IUPAC Name (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES C(C(C1C(C(C(=O)O1)O)O)O)O

Specifications

Melting Point 184°C to 190°C
Color White
Quantity 5 g
Beilstein 83002
Solubility Information Soluble in water
Optical Rotation +54.5° (c=4 in Water)
Formula Weight 178.14
Percent Purity 95%
Physical Form Powder
Chemical Name or Material L-Gulonic acid-1,4-lactone
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only