missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Glutamic acid monosodium salt

Catalog No. AAJ6342414
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6342414 25 g
AAJ6342409 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6342414 Supplier Thermo Scientific Chemicals Supplier No. J6342414

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 142-47-2 | C5H8NNaO4 | 169.11 g/mol

L-Glutamic acid monosodium salt is used as a flavor enhancer, swine feed additive as well as to improve the taste of bitter drugs. It is used therapeutically to reduce blood ammonia levels. It serves as an excitatory amino acid neurotransmitter, which is an agonist at glur and NMDA receptors. Further, it is used to reduce the intake of sodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water. Insoluble in ether.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 142-47-2
Molecular Formula C5H8NNaO4
Molecular Weight (g/mol) 169.11
MDL Number MFCD00150138
InChI Key LPUQAYUQRXPFSQ-UHFFFAOYNA-M
Synonym natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt
PubChem CID 86748263
SMILES [Na+].NC(CCC(O)=O)C([O-])=O

Specifications

Color White
Merck Index 14,6254
Solubility Information Soluble in water. Insoluble in ether.
IUPAC Name (2S)-2-aminopentanedioic acid;sodium
Quantity 25 g
Formula Weight 169.11
Physical Form Powder
Chemical Name or Material L-Glutamic acid monosodium salt
EINECSNumber 205-538-1
RTECSNumber MA1578000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only