missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid Monopotassium Salt, Spectrum™ Chemical

Catalog No. 18603829
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603829 2.5 kg Poly Bottle
18603827 100 g Poly Bottle
18603828 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603829 Supplier Spectrum Chemical Mfg Cor Supplier No. G10902.5KG

G1090, 6382-01-0, C5H8KNO4·H2O

Spectrum™ Chemical L-Glutamic Acid Monopotassium Salt , also known as sodium glutamate, is the sodium salt of glutamic acid. It is used in the food industry most usually as a flavor with an umami taste. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 6382-01-0
Molecular Formula C5H10KNO5
Molecular Weight (g/mol) 203.24
InChI Key XIBUKSQTWSKJMQ-QTNFYWBSSA-M
IUPAC Name potassium (4S)-4-amino-4-carboxybutanoate hydrate
SMILES O.[K+].N[C@@H](CCC([O-])=O)C(O)=O

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 2.5 kg
Grade Technical
Chemical Name or Material L-Glutamic Acid Monopotassium Salt