missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid monopotassium salt monohydrate, 97+%

Catalog No. AAA1723230
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1723230 250 g
AAA172320B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1723230 Supplier Thermo Scientific Chemicals Supplier No. A1723230
Only null left

CAS: 6382-01-0 | C5H11KNO5 | 204.243 g/mol

L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 6382-01-0
Molecular Formula C5H11KNO5
Molecular Weight (g/mol) 204.243
MDL Number MFCD00150343
InChI Key VMWJTBYKMYBHOC-QTNFYWBSSA-N
Synonym l-glutamic acid hydrate potassium, l-glutamicacid,monopotassiumsalt,monohydrate
PubChem CID 87237506
IUPAC Name (2S)-2-aminopentanedioic acid;potassium;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O.[K]

Specifications

Melting Point 115°C to 118°C
Solubility Information Soluble in water.
Quantity 250 g
Formula Weight 203.25 (185.23 Anhydrous)
Percent Purity ≥97%
Chemical Name or Material L-Glutamic acid monopotassium salt monohydrate
RTECSNumber MA1450000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only