missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid monopotassium salt monohydrate, 97+%

CAS: 6382-01-0 | C5H11KNO5 | 204.243 g/mol

Supplier:  Thermo Scientific Chemicals A172320B

Quantity 1000 g

 View more versions of this product

Catalog No. AAA172320B


Only null left

Description

Description

L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid monopotassium salt monohydrate acts as a common excitatory neurotransmitter like the agonist at kainate, N-methyl-D-aspartate receptor (NMDA) and quisqualate receptors in central nervous system. It is also used in cell culture as a component of MEM non-essential amino acids solution..

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

6382-01-0
204.243
VMWJTBYKMYBHOC-QTNFYWBSSA-N
87237506
C(CC(=O)O)C(C(=O)O)N.O.[K]
C5H11KNO5
MFCD00150343
l-glutamic acid hydrate potassium, l-glutamicacid,monopotassiumsalt,monohydrate
(2S)-2-aminopentanedioic acid;potassium;hydrate

Specifications

115°C to 118°C
6382-01-0
MFCD00150343
Soluble in water.
C(CC(=O)O)C(C(=O)O)N.O.[K]
1000 g
87237506
≥97%
L-Glutamic acid monopotassium salt monohydrate
C5H11KNO5
l-glutamic acid hydrate potassium, l-glutamicacid,monopotassiumsalt,monohydrate
VMWJTBYKMYBHOC-QTNFYWBSSA-N
(2S)-2-aminopentanedioic acid;potassium;hydrate
204.243
203.25 (185.23 Anhydrous)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MA1450000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions