missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, free acid, ≥99%, MP Biomedicals™

Catalog No. ICN10180091
Change view
Click to view available options
Quantity:
100 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10180091 1 kg
ICN10180080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10180091 Supplier MP Biomedicals Inc Supplier No. 0210180091

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-47-2
Molecular Formula C5H8NNaO4
Molecular Weight (g/mol) 169.11
MDL Number MFCD00150138
InChI Key LPUQAYUQRXPFSQ-UHFFFAOYNA-M
Synonym natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt
PubChem CID 86748263
IUPAC Name sodium 2-amino-4-carboxybutanoate
SMILES [Na+].NC(CCC(O)=O)C([O-])=O

Specifications

Color White
Assay Percent Range ≥99%
Density 1.538g/mL at 20°C
Quantity 1 kg
Formula Weight 187.13
Percent Purity ≥99%
Grade Research
Physical Form Powder
Chemical Name or Material L-Glutamic acid Monosodium Salt
EINECSNumber 205-538-1
Recommended Storage Store at room temperature (15 to 30°C).