missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid alpha-(7-amido-4-methylcoumarin)

Catalog No. AAJ64847MF
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ64847MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ64847MF Supplier Thermo Scientific Chemicals Supplier No. J64847MF
Only null left

CAS: 98516-76-8 | C15H16N2O5 | 304.302 g/mol

A fluorogenic substrate for aminopeptidase a, producing a blue fluorescent solution upon cleavage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A fluorogenic substrate for aminopeptidase a, producing a blue fluorescent solution upon cleavage.

Chemical Identifiers

CAS 98516-76-8
Molecular Formula C15H16N2O5
Molecular Weight (g/mol) 304.302
MDL Number MFCD00051217
InChI Key USALUSONPMPEKY-NSHDSACASA-N
Synonym l-glutamic acid alpha-7-amido-4-methylcoumarin, glu-nh-med, 4s-4-amino-5-4-methyl-2-oxochromen-7-yl amino-5-oxopentanoic acid, s-4-amino-5-4-methyl-2-oxo-2h-chromen-7-yl amino-5-oxopentanoic acid, 4s-4-amino-4-4-methyl-2-oxochromen-7-yl carbamoyl butanoic acid, ambotzhaa7670, 7-alpha-glutamyl-4-methylcoumarylamide, h-glu-amc, alpha-glutamyl-7-amino-4-methylcoumarylamide, n-4-methyl-2-oxo-2h-chromen-7-yl-l-, a-glutamine
PubChem CID 126959
IUPAC Name (4S)-4-amino-5-[(4-methyl-2-oxochromen-7-yl)amino]-5-oxopentanoic acid
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)NC(=O)C(CCC(=O)O)N

Specifications

Color White
Quantity 50 mg
Formula Weight 304.3
Physical Form Powder
Chemical Name or Material L-Glutamic acid alpha-(7-amido-4-methylcoumarin)
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only