missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure

Catalog No. AAJ16215C4
Change view
Click to view available options
Quantity:
1 kg
25 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ16215C4 25 kg Plastic Bottle
AAJ16215A1 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ16215C4 Supplier Thermo Fisher Scientific Supplier No. J16215C4
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-VKHMYHEASA-N
Synonym l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex
PubChem CID 33032
ChEBI CHEBI:16015
IUPAC Name (2 S)-2-aminopentanedioic acid
SMILES C(CC(=O)O)C(C(=O)O)N

Specifications

Melting Point 189°C to 192°C (decomposition)
Packaging Plastic Bottle
Linear Formula HOOCCH2CH2CH(NH2)COOH
Beilstein 1723801
Density 1.525
Quantity 25 kg
Formula Weight 147.13
Percent Purity 98.5 to 101.5%
Specific Gravity 1.525
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material L-Glutamic acid
Show More Show Less
Hazard Category H500
EINECSNumber 200-293-7
RTECSNumber LZ9700000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only