missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 1-methyl ester, 98%

Catalog No. AAH6327906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6327906 5 g
AAH6327903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6327906 Supplier Thermo Scientific Chemicals Supplier No. H6327906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6384-08-3 | C8H12N2 | 136.20 g/mol

L-Glutamic acid 1-methyl ester nonessential amino acid that is important to the mammalian central nervous system. It is also a neurotransmitter for cone photoreceptors in the human brain and is used as a treatment for patients who have liver disease accompanied by encephalopathy (a condition termed ‘hepatic coma’).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.

Chemical Identifiers

CAS 6384-08-3
MDL Number MFCD00171677

Specifications

Melting Point 145°C (decomposition)
Color White
Molecular Formula C6H11NO4
Beilstein 1707273
Synonym h-glu-ome, s-4-amino-5-methoxy-5-oxopentanoic acid, l-glutamic acid 1-methyl ester, l-glutamate methylester, 1-methyl l-glutamate, l-glutamate methyl ester, glutamic acid methyl ester, 4s-4-amino-5-methoxy-5-oxopentanoic acid, gluome, glu alpha-ome-oh
Solubility Information Soluble in water.
InChI Key SEWIYICDCVPBEW-UHFFFAOYNA-N
SMILES COC(=O)C(CCC(=O)O)N
IUPAC Name (4S)-4-amino-5-methoxy-5-oxopentanoic acid
Quantity 5 g
Molecular Weight (g/mol) 161.16
PubChem CID 440550
ChEBI CHEBI:28575
Formula Weight 161.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Glutamic acid 1-methyl ester
Show More Show Less
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only