missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-(-)-Fucose, ≥95.0% (GC), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 2438-80-4 |
| Linear Formula | C6H12O5 |
| MDL Number | MFCD00135607 |
| Melting Point | 150°C to 153°C |
| Synonym | 6-Deoxy-L-galactose |
| Type | Analytical Standard |
| Quantity | 25 mg |
| Formula Weight | 164.16 |
| Percent Purity | ≥95.0% (GC) |
| Grade | Analytical Standard |