missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine Dimethyl Ester Dihydrochloride 96.0+%, TCI America™

Catalog No. p-7125919
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C33035G 5 g
C330325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C33035G Supplier TCI America Supplier No. C33035G

Chemical Identifiers

CAS 32854-09-4
Molecular Formula C8H18Cl2N2O4S2
Molecular Weight (g/mol) 341.26
MDL Number MFCD00012490,MFCD00012490,MFCD09951837,MFCD00012490
InChI Key QKWGUPFPCRKKMQ-UHFFFAOYNA-N
Synonym h-cys-ome 2.2hcl, dimethyl l-cystinate dihydrochloride, h-d-cys-ome 2 2hcl, methyl 2-amino-3-r-2-amino-3-methoxy-3-oxopropyl disulfanyl propanoate dihydrochloride, d-cystine bis methyl ester dihydrochloride, d-cystine dimethyl ester dihydrochloride, h-d-cys-ome 2 hcl, c8h16n2o4s2.2hcl, dimethyl l-cystinate 2hcl, h-d-cys-ome 2 . 2 hcl
PubChem CID 71463656
IUPAC Name dihydrogen methyl 2-amino-3-[(2-amino-3-methoxy-3-oxopropyl)disulfanyl]propanoate dichloride
SMILES [H+].[H+].[Cl-].[Cl-].COC(=O)C(N)CSSCC(N)C(=O)OC

Specifications

Melting Point 179°C
Color White
Quantity 5 g
Formula Weight 341.26
Percent Purity ≥96.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Cystine Dimethyl Ester Dihydrochloride