missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Cysteine hydrochloride, anhydrous, 98%

Catalog No. AAL0632814
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0632814 25 g
AAL0632822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0632814 Supplier Thermo Scientific Chemicals Supplier No. L0632814

L-cysteine hydrochloride , CAS# 7048-04-6, also known as L-cysteine hydrochloride monohydrate, is a soluble salt of the non-essential amino acid, L-cysteine. It is commonly used as a nucleophile in several chemical reactions. | CAS: 52-89-1 | C3H8ClNO2S | 157.61 g/mol

L-Cysteine hydrochloride, anhydrous is widely used as additive in food production. It is used as an antioxidant to promote fermentation and keep the flavor. It is also used in cosmetics and animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-cysteine hydrochloride is a non-essential proteinogenic amino acid commonly used in chemical reactions, for instance, the thiol side chain in cysteine often participates in enzymatic reactions as a nucleophile.

Application

  • L-cysteine hydrochloride plays an important role as a histidine ammonia-lyase inhibitor with the ability to interfere with the action of histidine ammonia-lyase.
  • In chemical reactions, L-cysteine hydrochloride is used as a raw material for the production of S-carboxymethyl-L-cysteine, L-cysteine base, and N-acetyl-L-cysteine.
  • This compound also has antioxidant activity.
  • It has been used in laboratory protocols to separate dorsal root ganglion neurons.
  • This compound is applied to study the inhibitory effect of L-cysteine hydrochloride hydrate in preventing enzymatic browning in apples.

Chemical Identifiers

CAS 52-89-1
Molecular Formula C3H8ClNO2S
Molecular Weight (g/mol) 157.61
MDL Number MFCD00064553
InChI Key IFQSXNOEEPCSLW-QYLAHFPANA-N
Synonym l-cysteine hydrochloride, cystein chloride, r-cysteine hydrochloride, l-cysteine.hcl, cysteine monohydrochloride, l-+-cysteine hydrochloride, l-cysteine, hydrochloride, l-cysteine monohydrochloride, cysteine hcl, l-cysteine hydrochloride anhydrous
PubChem CID 60960
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid;hydrochloride
SMILES Cl.N[C@@H](CS)C(O)=O

Specifications

Melting Point ∽180°C (decomposition)
pH 0.8 to 1.2
Beilstein 3560277
Merck Index 14,2781
Solubility Information Soluble in water. (580g/L at 20°C),
Quantity 25 g
Odor Irritating
Formula Weight 157.62
Optical Rotation +6.4° (c=5 in 5N HCl)
Percent Purity 98%
Sensitivity Air sensitive; Hygroscopic
Chemical Name or Material L-Cysteine hydrochloride, anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-157-7
RTECSNumber HA2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only