missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Carvone, 99%

Catalog No. AC154590050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC154590050 5 g Glass bottle
AC154591000 100 g Glass bottle
AC154595000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC154590050 Supplier Thermo Scientific Chemicals Supplier No. 154590050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6485-40-1 | C10H14O | 150.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6485-40-1
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.22
MDL Number MFCD00001578
InChI Key ULDHMXUKGWMISQ-SECBINFHSA-N
Synonym --carvone, l-carvone, r---carvone, l--carvone, r-2-methyl-5-prop-1-en-2-yl cyclohex-2-enone, 4r-carvone, levo-carvone, --p-mentha-6,8-dien-2-one, --r-carvone
PubChem CID 439570
ChEBI CHEBI:15400
IUPAC Name (5R)-2-methyl-5-prop-1-en-2-ylcyclohex-2-en-1-one
SMILES CC(=C)[C@@H]1CC=C(C)C(=O)C1

Specifications

Color Colorless to Yellow
Density 0.9580g/mL
Boiling Point 227.0°C to 230.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4950 to 1.5020
Quantity 5 g
Beilstein 07, 157
Specific Gravity 0.958
Merck Index 14, 1874
Specific Rotation − 57.00
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in alcohol, ether, chloroform,, propylene glycol and mineral oils, insoluble in glycerol
Formula Weight 150.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material L(-)-Carvone, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing.
EINECSNumber 229-352-5
RTECSNumber OS8650000
TSCA TSCA

RUO – Research Use Only