missing translation for 'onlineSavingsMsg'
Learn More

L-Carnitine L-Tartrate 98.0+%, TCI America™

Numéro de catalogue. p-7125564
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
C26255G 5 g
C262525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. C26255G Fournisseur TCI America Code fournisseur C26255G
Il en reste null

Identifiants chimiques

CAS 36687-82-8
Molecular Formula C11H21NO9
Molecular Weight (g/mol) 311.29
MDL Number MFCD04974128
InChI Key RZALONVQKUWRRY-HMWYIXBMNA-N
Synonym Bis[[(R)-3-carboxy-2-hydroxypropyl]trimethylammonium] L-Tartrate

Spécifications

Color White
Quantity 5 g
Formula Weight 236.24
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Carnitine L-Tartrate