missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid dimethyl ester hydrochloride, 98%

Catalog No. AC376660250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC376660250 25 g Plastic Bottle
AC376660050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC376660250 Supplier Thermo Scientific Chemicals Supplier No. 376660250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 32213-95-9 | C6H12ClNO4 | 197.615 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32213-95-9
Molecular Formula C6H12ClNO4
Molecular Weight (g/mol) 197.615
MDL Number MFCD00038878
InChI Key PNLXWGDXZOYUKB-WCCKRBBISA-N
Synonym h-asp ome-ome hcl, l-aspartic acid dimethyl ester hydrochloride, dimethyl l-aspartate hydrochloride, h-asp ome-ome.hcl, methyl aspartic acid hydrochloride, s-dimethyl 2-aminosuccinate hydrochloride, aspartic acid dimethyl ester hydrochloride, l-aspartic acid dimethyl ester hydro-chloride, s-aminosuccinic acid dimethyl ester hydrochloride, dimethyl aspartic acid hydrochloride
PubChem CID 2734892
IUPAC Name dimethyl (2S)-2-aminobutanedioate;hydrochloride
SMILES COC(=O)CC(C(=O)OC)N.Cl

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic Bottle
Linear Formula CH3OC(O)CH2CH(NH2)C(O)OCH3·HCl
Specific Rotation +13° to +17° (20°C, 589nm) (c=1, CH3OH)
Specific Rotation Condition +15° (20°C c=1,Chloroform)
Quantity 25 g
Formula Weight 197.62
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Aspartic acid dimethyl ester hydrochloride
Show More Show Less

RUO – Research Use Only