missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid beta-benzyl ester, 98%

Catalog No. AC441930250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC441930250 25 g
AC441930050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC441930250 Supplier Thermo Scientific Chemicals Supplier No. 441930250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2177-63-1 | C11H13NO4 | 223.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2177-63-1
Molecular Formula C11H13NO4
Molecular Weight (g/mol) 223.23
InChI Key VGALFAWDSNRXJK-VIFPVBQESA-N
Synonym h-asp obzl-oh, l-aspartic acid 4-benzyl ester, l-aspartic acid beta-benzyl ester, beta-benzyl l-aspartate, 2s-2-amino-4-benzyloxy-4-oxobutanoic acid, 4-benzyl l-aspartate, benzyl hydrogen beta-l-aspartate, s-2-amino-4-benzyloxy-4-oxobutanoic acid, 2s-2-amino-4-oxo-4-phenylmethoxybutanoic acid, asp obzl
PubChem CID 101186
IUPAC Name (2S)-2-amino-4-oxo-4-phenylmethoxybutanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CC(C(=O)O)N

Specifications

Melting Point 207°C to 208°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 25 g
Specific Rotation + 27.8
Formula Weight 223.23
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Aspartic acid β-benzyl ester
Show More Show Less
EINECSNumber 218-541-8

RUO – Research Use Only