missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride, 98%

Catalog No. p-7056807
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6593703 1 g
AAH6593706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6593703 Supplier Thermo Scientific Chemicals Supplier No. H6593703

CAS: 94347-11-2 | C15H22ClNO4 | 315.79 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94347-11-2
Molecular Formula C15H22ClNO4
Molecular Weight (g/mol) 315.79
MDL Number MFCD00190761
InChI Key YSDLFDMXCRFHTQ-UHFFFAOYNA-N
Synonym s-4-benzyl 1-tert-butyl 2-aminosuccinate hydrochloride, h-asp obzl-otbu.hcl, 4-benzyl 1-tert-butyl 2s-2-aminobutanedioate hydrochloride, c15h21no4.hcl, h-asp obzl-otbu hcl, h-asp obzl-otbu hydrochloride, s-4-benzyl 1-tert-butyl 2-aminosuccinate hcl, l-aspartic acid beta-benzyl ester alfa-t-butyl ester hcl, 4-benzyl 1-tert-butyl s-2-aminosuccinate hydrochloride, 4-benzyl 1-tert-butyl l-aspartate-hydrogen chloride 1/1
PubChem CID 56777343
IUPAC Name 4-benzyl 1-tert-butyl 2-aminobutanedioate hydrochloride
SMILES Cl.CC(C)(C)OC(=O)C(N)CC(=O)OCC1=CC=CC=C1

Specifications

Color White
Beilstein 2538326
Quantity 1 g
Formula Weight 315.79
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Aspartic acid 4-benzyl ester 1-tert-butyl ester hydrochloride
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only