missing translation for 'onlineSavingsMsg'
Learn More

L-Aspartic acid 1-methyl ester, 98%

Catalog No. AAH6322106
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6322106 5 g
AAH6322103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6322106 Supplier Thermo Scientific Chemicals Supplier No. H6322106
Only null left

CAS: 17812-32-7 | C5H9NO4 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 17812-32-7
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00080289
InChI Key SWWBMHIMADRNIK-UHFFFAOYNA-N
Synonym h-asp-ome, 1-methyl l-aspartate, l-aspartic acid 1-methyl ester, s-3-amino-4-methoxy-4-oxobutanoic acid, 3s-3-amino-4-methoxy-4-oxobutanoic acid, l-aspartic acid methyl ester, pubchem13176, l-aspartic acid 1-methyl ester; 3-amino-4-methoxy-4-oxobutanoic acid; h-asp-ome
PubChem CID 6994682
SMILES COC(=O)C(N)CC(O)=O

Specifications

Melting Point 186°C (decomposition)
Color White
Beilstein 2080763
IUPAC Name (3S)-3-amino-4-methoxy-4-oxobutanoic acid
Quantity 5 g
Formula Weight 147.13
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Aspartic acid 1-methyl ester
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only