missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L(+)-Asparagine monohydrate, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécifications
Spécifications
| Melting Point | 233.0°C to 235.0°C |
| Ammonium | 1000ppm max. |
| Arsenic (As) | 1ppm max. |
| Color | White |
| Chlorine (Cl) | 210ppm max. |
| Heavy Metals (as Pb) | 10ppm max. |
| Infrared Spectrum | Authentic |
| Iron (Fe) | 10ppm max. |
| Loss on Drying | 11.5% to 12.5% (130°C, 3 hrs) |
| Assay Percent Range | 99% |
| Afficher plus de résultats |
RUO – Research Use Only