missing translation for 'onlineSavingsMsg'
Learn More

L-Asparagine anhydrous, 100%, MP Biomedicals™

Catalog No. ICN10079425
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10079425 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN10079425 Supplier MP Biomedicals Inc Supplier No. 0210079425

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Anhydrous cell culture reagent provided as a white powder suitable for research purpose

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Color White
Assay Percent Range 1
Quantity 25 g
Formula Weight 132.1
Grade Cell Culture
Physical Form Powder
Chemical Name or Material L-Asparagine