missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-(+)-Asparagine, 99%

Catalog No. AAB2147336
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2147336 500 g
AAB2147322 100 g
AAB214730E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2147336 Supplier Thermo Scientific Chemicals Supplier No. B2147336

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 70-47-3 | C4H8N2O3 | 132.119 g/mol

L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(+)-Asparagine acts as a precursor to aspartate. It is also used as a diuretic. It is also employed in the biosynthesis of glycoproteins and proteins. It acts as a food additive. It reacts with reducing sugar to yield acrylamide, which finds application in french fries, potato chips, and toasted bread. It plays an important role in the function and development of the brain. Further, it is useful for human body, physically and mentally. In addition to this, it is used in cell culture media and is a component of (2-methoxyethoxy)methyl (MEM) non-essential amino acid solution.

Solubility
Soluble in acids, bases. Insoluble in methanol, ethanol, ether and benzene.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 70-47-3
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.119
MDL Number MFCD00064401
InChI Key DCXYFEDJOCDNAF-REOHCLBHSA-N
Synonym l-asparagine, asparagine, s-asparagine, aspartamic acid, agedoite, altheine, h-asn-oh, crystal vi, asparamide, asparagine acid
PubChem CID 6267
ChEBI CHEBI:17196
IUPAC Name (2S)-2,4-diamino-4-oxobutanoic acid
SMILES C(C(C(=O)O)N)C(=O)N

Specifications

Melting Point 234°C to 236°C (decomposition)
Beilstein 1723527
Merck Index 14,837
Solubility Information Soluble in acids,bases. Insoluble in methanol,ethanol,ether and benzene.
Quantity 500 g
Formula Weight 132.12
Percent Purity 99%
Sensitivity Hygroscopic
Chemical Name or Material L-(+)-Asparagine
EINECSNumber 200-735-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only