missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ascorbyl palmitate, specified according to requirements of USP/Ph.Eur., Thermo Scientific Chemicals

Catalog No. AC449481000
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC449481000 100 g
AC449480250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC449481000 Supplier Thermo Scientific Chemicals Supplier No. 449481000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 137-66-6
Molecular Formula C22H38O7
Molecular Weight (g/mol) 414.54
MDL Number MFCD00005377
InChI Key NULLRXHRYOXSEW-UHFFFAOYNA-N
Synonym ascorbyl palmitate, l-ascorbyl 6-palmitate, 6-o-palmitoyl-l-ascorbic acid, ascorbic palmitate, l-ascorbyl palmitate, unii-qn83us2b0n, ascorbyl monopalmitate, l-ascorbic acid 6-palmitate, s-2-r-3,4-dihydroxy-5-oxo-2,5-dihydrofuran-2-yl-2-hydroxyethyl palmitate, ascorbyl palmitate nf
PubChem CID 54680660
IUPAC Name [(2S)-2-[(2R)-3,4-dihydroxy-5-oxo-2H-furan-2-yl]-2-hydroxyethyl] hexadecanoate
SMILES CCCCCCCCCCCCCCCC(=O)OCC(O)C1OC(O)=C(O)C1=O

Specifications

Melting Point 111°C to 115°C
Boiling Point 250°C
Ignition Residue 0.1% max
Packaging Glass bottle
Quantity 100 g
Solubility Information Solubility in water: insoluble
Formula Weight 414.53
Chemical Name or Material L(+)-Ascorbyl palmitate
EINECSNumber 205-305-4

RUO – Research Use Only